Tlačiť
Precast gels, ExcelGel
Dodávateľ: Cytiva
17-6002-21EA 0 EUR
GEHE17-6002-21 GEHE17-1236-01
Precast gels, ExcelGel
Precast polyacrylamide gels make a wide range of electrophoretic techniques readily available with minimum preparation. Manufacturing quality and reproducibility of these gels translate into highly reliable and reproducible results with minimal training.
  • ExcelGel precast SDS polyacrylamide gels available as 12,5% homogeneous or 12% to 14% gradient gels
ExcelGel precast SDS polyacrylamide gels available as 12,5% homogeneous or 12% to 14% gradient gels.