Tlačiť
Water purification systems, Direct-Q® 5 UV, Direct-Q® 8 UV, Millipore®
Dodávateľ: Merck
Direct-Q®
171-0819 171-0818 171-0835 171-0833 171-0834 171-0831 171-0798 171-0799 171-0832 171-0796 171-0830 171-0797 171-0794 171-0795 171-0792 171-0793 171-0790 171-0791 171-0808 171-0828 171-0806 171-0807 171-0829 171-0804 171-0826 171-0805 171-0827 171-0824 171-0825 171-0822 171-0789 171-0823 171-0820 171-0787 171-0788 171-0821
Water purification systems, Direct-Q® 5 UV, Direct-Q® 8 UV, Millipore®
Systémy pro výrobu čisté vody Systémy pro výrobu čisté a ultračisté vody
The Direct-Q® range of systems provides the user with a convenient and flexible solution for pure and ultrapure water requirements, directly from potable tap water. The user will have access to ultrapure water for critical applications, and pure water for less critical applications such as general glassware washing or final rinsing from the same system. The high quality ultrapure water produced by Direct-Q® systems is suitable for applications such as production of mobile phase for chromatographic separations; preparation of blanks and standard solutions for spectrophotometry, spectroscopy or other analytical techniques; and preparation of buffers for biochemical experiments. For the production of type I and type III water.
  • An integral timer makes it possible to dispense type I water for a specified time period; the system switches off automatically when the set time has elapsed
  • An alarm is triggered when there is a sudden drop in the quality of the product water
  • Water quality parameters (resistivity or conductivity, temperature) are shown on a colour display
  • Maintenance consists of a simple pack replacement once or twice a year
Designed to fit perfectly into the lab environment, the versatile remote dispenser can be placed up to two metres from Direct-Q® water purification unit. The units can be bench or wall mounted (wall mount bracket required). Alternatively, Direct-Q® systems are also available with an integrated dispenser for use on the bench top. The design of the bench-installed Direct-Q® systems is also adapted to the height and shape of common laboratory glassware.
Direct-Q® systems include with a built-in 185/254 nm UV lamp to reduce the level of organics for critical applications. Water with low TOC provides important benefits to HPLC users such as higher sensitivity and longer column lifetime. The same UV lamp also destroys bacteria.
Pure (type III) product water quality values (in regular operating conditions):
Ionic rejection >96%
Organic rejection for MW >200 >99%
Bacteria and particulates rejection >99%
Objednávacie informácie: SmartPak™ should be added with a Millipak® Express or a BioPak® final filter.
Objednávacie informácie: Delivered with main Direct-Q® system. Please order Millipak® Express or a BioPak® final filter, tank vent filter and SmartPak™ DQ3 (for Direct-Q® 5 UV) or DQ8 (for Direct-Q® 8 UV) cartridge separately.
Pozor: Particulates, bacteria, endotoxins (pyrogens), RNases, DNases levels quoted are with Millipak® Express or with BioPak® as final polisher.