Tlačiť
Ice makers, FM series
Dodávateľ: Hoshizaki
474-0136 474-0118
Ice makers, FM series
Výrobníky ledu
The FM series produce flake or nugget ice, with the nugget ice compressed during production to make it long-lasting. The flake and nugget ice is popular for a variety of cooling purposes including the display of fish and fresh produce, and also in the medical world for cooling blood and organs.
  • Automatic flush function: regular flushing to keep the ice making system clean
  • One piece bin moulding: round corners are hygienic and easy to keep clean
  • Integrated door handle design: easy to open and to clean
  • Controller board with operation status display: ensuring product durability and low maintenance costs