Tlačiť
Sterilisation indicator tapes
Dodávateľ: Deltalab
DELT191050EA 349.44 EUR
DELT191050
Sterilisation indicator tapes
Indicator for sterilisation by ethylene oxide gas.
  • Suitable for sticking on different surfaces such as plastic, metal, fabrics, paper and cardboard
  • Tape will remain securely affixed for 18 months
The printed letters GAS change colour from red to green by effect of time, temperature, humidity and EO concentration as per the exposition to 500 mg/L EO and 60% RH for minimum 30 min at 30 °C, or exposition to 12% EO and 60% RH for minimum 48 min at 54 °C.