Chcete sa opäť prihlásiť?
Pestles, KONTES®, PELLET PESTLES®
Dodávateľ: KIMBLE/KONTES
KONTES® PELLET PESTLE®
Grinders are ideal for resuspending protein and DNA pellets or grinding soft tissues. Pestles are specially designed to fit either 0,5 ml or 1,5 ml microtubes.
Disposable, autoclavable polypropylene pestles eliminate the need for pretreatment of pestles and microtubes. The CTFE/stainless steel pestles are reusable.
Ordering information: Pestle may be hand-operated, or driven by optional cordless mixer motor. Mixer motor includes two AA size batteries and adapter for attaching pestle.
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Dĺžka Pk Číslo v katalógu VWR Množstvo
Pistils, polypropylene, 100 pcs 70 100 432-1853 Unavailable
unavailable Tento produkt už nie je dostupný. Môžu existovať alternatívy, ktoré sa dajú vyhľadať pomocou čísla VWR katalógu, ktoré je uvedené vyššie. Ak potrebujete pomoc, prosím zavolajte VWR služby zákazníkom na +43 1 97002 - 0.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo