Tlačiť
Tubing for vacuum applications, Tygon® S3™ E-3603
Dodávateľ: Saint-Gobain
Tygon®
228-1361EA 2650.04 EUR
228-1361 228-1360 228-1362 228-1264 228-1266
Tubing for vacuum applications, Tygon® S3™ E-3603
Hadičky Hadičky Tygon
Non-DEHP tubing, extra heavy walls vacuum applications.
  • Outstanding chemical resistance
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Lot-to-lot consistency for reproducible results
Tubing handles most inorganic chemicals in the laboratory. It is non-oxidising and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning, and coils are marked at I-foot (30,4 cm) intervals for easy measuring. This thick-walled tubing withstands vacuum up to 759 mm of mercury at 23 °C and 686 mm of mercury at 60 °C.
Temperature resistance: −46 to +74 °C
Hardness (Shore durometer A): 56
Density: 1,18
Certifikácie: Complies with FDA, NSF 51 & 61, meets USP Class VI