Tlačiť
Disinfectant wipes, Mikrobac® Tissues / Mikrobac® Tissues XXL
Dodávateľ: Bode
Mikrobac® Tissues
BODL977040 115-2159 BODL981339
Disinfectant wipes, Mikrobac® Tissues / Mikrobac® Tissues XXL
Ubousky/Utěrky Utěrky jednorázové a ručníky Ubrousky dezinfekční
These aldehyde, alcohol, fragrance and colourant-free rapid disinfectant wipes are ideal for simple disinfection of alcohol sensitive surfaces (e.g. in clinics, labs and medical practices) and medical devices. They are also suitable for the disinfectant cleaning of non-submersible ultrasonic transducers or probes.
  • Especially gentle on materials
  • Robust PET fibre material shows no adsorption and fully delivers active substances
  • Safe and easy dispensing of individual wipes from resealable flowpack
  • Disinfected surfaces can be used again after an exposure time of just 1 minute
Mikrobac® Tissues are ideal for the disinfection of smaller surfaces (approx. 1 m²) while Mikrobac® Tissues XXL can be used for larger surfaces (approx. 2 m²).
Microbiological effectiveness: bactericidal, yeasticidal, limited virucidal action (incl. HBV, HIV, HCV), and on MNV, polyoma- and rotaviruses.
In order to avoid premature drying out of the wipes, please reseal the pack immediately after use.
Balenie: 40 or 80 wipes per resealable, handy flowpack. A wall mounting for the pack is also available.
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904