Tlačiť
Protein quantitation assay kits, SPN™ Protein assay
Dodávateľ: G-Biosciences
786-020EA 568.18 EUR
GENO786-020 GENO786-021
Protein quantitation assay kits, SPN™ Protein assay
A spin column format protein assay that is suitable for single sample or high throughput protein estimation. The SPN™ and SPN™-htp protein assays are rapid assays that are suitable for as little as 0,5 µg protein and are resistant to interference from common laboratory agents. The assays are excellent for the rapid determination of protein samples in Laemmli or other SDS-PAGE loading buffers.