Tlačiť
Hybridisation bath, Slide Moat™
Dodávateľ: Boekel Scientific
Boekel
731-0524 731-0526
Hybridisation bath, Slide Moat™
Sušárny Pece hybridizační
The Slide Moat™ is a versatile microscope slide incubator designed for in situ hybridisation or denaturation, and is ideal for all slide drying and warming procedures used in histology, cytology or pathology laboratories.
  • 30 standard microscope slide capacity
  • PID temperature controller for easy temperature selection, rapid heat up and excellent temperature stability
  • Tight fitting, tempered glass lid, which seals to an integral gasket on the heating base
  • Airtight lid seals in moisture and only a few drops of water are required to maintain a saturated atmosphere for overnight incubations
Note: It is strongly recommended that covered and sealed slides are used for high temperature (>37 °C) or prolonged period (>1 hour) incubations.
Informace o dodávce: The Slide Moat™ bath is supplied complete with a tempered glass lid.