Tlačiť
Slide hybridisation oven, InSlide Out™
Dodávateľ: Boekel Scientific
Boekel
731-0529 731-0527 731-0528
Slide hybridisation oven, InSlide Out™
Sušárny Pece hybridizační
The InSlide Out™ hybridisation oven is designed for a variety of hybridisation and immunohistochemical procedures involving slides.
  • 20 slide capacity
  • Prepared slides are placed into the sealed aluminium tray for incubation, which maintains humidity and prevents the slides from drying out
  • The rapid heat up and excellent temperature stability ensure consistency from test to test
  • Optional PC slide holder insert accommodates up to 18 slides and may be placed directly into a water bath for wash procedures
Informace o dodávce: The InSlide Out™ oven is supplied complete with a sealed aluminium tray.