Tlačiť
Bovine Serum Albumin (BSA), 20 mg/ml in aqueous solution, for biotechnology
J642-1MLEA 0 EUR
VWRCJ642-1ML VWRCJ642-5ML
Bovine Serum Albumin (BSA), 20 mg/ml in aqueous solution, for biotechnology
Contains 0,1% sodium azide as preservative and strictly tested for RNase, DNase and protease making it suitable for sensitive applications.