Tlačiť
Industrial gloves, oil repellent, HyFlex® 11-927
Dodávateľ: Ansell
HyFlex®
112-4301EA 343.76 EUR
112-4301 112-4300 112-4299 112-4298 112-4297 112-4296
Industrial gloves, oil repellent, HyFlex® 11-927
Rukavice
These flexible gloves combine advanced cut resistance, oil repellence and oil grip. Made of nylon/HPPE knitting with nitrile coating they are ideal for handling sharp metal components coated in oil or lubricant. Also suitable for assembling applications, metal working and maintenance tasks.
  • ¾ nitrile dip provides extra protection against oil exposure and knuckle abrasion
  • Ansell Grip™ technology uses microscopic channels to wick away oil and moisture from part surfaces and tools, providing a good grip
  • Shape offers superior fit and enables more agility in medium-duty applications
Certifikácie: EN 388:2016 (4342B), Cat. II