Tlačiť
Protective gel lotions, Stokoderm® Glove & Grip
Dodávateľ: SC Johnson
Stokoderm® Glove & Grip
115-2099
Protective gel lotions, Stokoderm® Glove & Grip
Péče o pokožku Krémy pro pokožku Vody pleťová na ruce a tělo
Gel-lotion helps to protect the skin from softening under gloves. Typical hazards: Wearing gloves, water-based workplace substances or light mechanical stress from e. g. glass fibre.
  • Increases the strength of the upper skin layer
  • Absorbs quickly and conomical in use
  • Contains BIO-ingredients and natural protecting complex
  • Ivory-coloured gel-lotion (O/W emulsion) perfumed or perfume-free
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904