Tlačiť
Cassettes for pump tubings
Dodávateľ: Ismatec
224-1981EA 95.51 EUR
224-1981 ISMC78001-95 ISMCIS0123A
Cassettes for pump tubings
Tlakové kazety
Click ’n’ go cassettes are suitable for non-monitored, long-time use, and provides automatic tubing pressure, and no adjustment necessary.
They are not suitable for differential pressure greater than 1 bar, for these conditions pressure lever cassettes are needed. Pressure lever cassettes allows to set a different tubing pressure for each channel.