Tlačiť
Pump Tubing, Tygon® LMT-55
Dodávateľ: Ismatec
Tygon®
ISMC06507-15EA 229.22 EUR
ISMC06507-15 ISMCSC0043T ISMC05502-26 ISMC05502-29 ISMCSC0226T ISMCSC0380T ISMCSC0048T ISMCSC0534T ISMC97618-26 ISMC97618-10 ISMCSC0027T ISMC97618-09 ISMCSC0064T ISMCSC0067T ISMC97619-36 ISMC97618-30 ISMC06507-39 ISMC97619-18 ISMCSC0047T ISMCSC0030T ISMC97618-44 ISMCSC0059T ISMCSC0072T ISMCSC0052T ISMCSC0002T ISMC97619-30 ISMC97618-16 ISMCSC0028T ISMC06507-05 ISMC97618-22 ISMC97619-44 ISMCSC0041T ISMCSC0502T ISMC97618-31 ISMCSC0039T ISMC97618-27 ISMC97619-31 ISMCSC0035T ISMCSC0058T ISMCSC0049T ISMCSC0694T ISMCSC0040T ISMCSC0068T ISMC97618-14 ISMCSC0057T ISMC06507-43 ISMCSC0006T ISMCSC0189T ISMCSC0224T ISMCSC0050T ISMCSC0054T ISMCSC0055T ISMCSC0503T ISMCSC0037T ISMC06507-11 ISMCSC0063T ISMC97618-18 ISMCSC0032T ISMC97619-29 ISMCSC0535T ISMC97619-24 ISMCSC0045T ISMC97618-34 ISMCSC0042T ISMC97618-46 ISMC05502-10 ISMCSC0691T ISMCSC0034T ISMCSC0376T ISMC97618-29 ISMCSC0504T ISMCSC0044T ISMCSC0071T ISMC97618-36 ISMC06507-09 ISMCSC0031T ISMCSC0051T ISMC97618-42 ISMCSC0029T ISMC97618-38 ISMC97618-28 ISMCSC0069T ISMC97618-39 ISMC97618-40 ISMCSC0223T ISMC05502-31 ISMCSC0046T ISMC97618-32 ISMCSC0026T
Pump Tubing, Tygon® LMT-55
Hadičky Hadičky pro pumpy
Tubing, non-DEHP and phthalate-free for laboratory applications. Crystal clear and flexible, long-lasting and crack-resistant, the non-DEHP tubing delivers the same superior performance you have come to expect, but now in a formulation that contains a non-DEHP plasticiser.
  • Outstanding chemical resistance
  • Lot-to-Lot consistency for reproducible results
  • Contains no BPA or phthalates
  • Temperature resistance:–50 to +74 °C
  • Hardness (Shore durometer A): 55
  • Density: 1,18
Handles most inorganic chemicals found in laboratory. It is non-oxidising and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning. Tubing is specially formulated for resistance to flex-fatigue and abrasion.
Length 381 mm, distance between stops 152 mm.
Length 381 mm, distance to stop 114 mm, distance between stops 72 mm (*).
Certifikácie: Complies with FDA, NSF, 3-A, meets USP Class VI