Tlačiť
Matrix needle printer
Dodávateľ: Kern & Sohn
KERN911-013
Matrix needle printer
Tiskárny Document Printers
Matrix needle printer to print the weights on normal paper which is ideal for long-term archiving.
  • Paper strips: 57 mm wide
Informace o dodávce: Supplied with interface cable RS 232.