Tlačiť
Bath fluids, Aquaclean
Dodávateľ: Hecht
HECH43170001 HECH43170003
Bath fluids, Aquaclean
Kapaliny do termostatu
Additive for keeping water bath and water circulating systems clean.
  • Prevents algae and bacterial growth
  • Prevents odour formation and obstructions in tubing and temperature control system
  • Saving of time: frequent cleaning of the tank is unnecessary
  • Solution is without aggressive phosphates and oxidising or reducing substances
  • Prevents foam formation, even with strong circulation
Pozor: Use biocides safely. Always read the label and product information before use.