Tlačiť
Emergency body shower, for wall mounting, modular
Dodávateľ: Broen Lab
118-0412EA 1309.37 EUR
118-0412
Emergency body shower, for wall mounting, modular
Pomůcky první pomoci Sprchy
Modular body shower for wall-mounting. In brass and stainless steel with chemical resistant red polycoating. With self-draining shower head for flexible wall mounting with pull rod.
  • With self-draining shower head for flexible wall mounting
  • With pull rod
  • Pre-mounted valve
  • Easy to mount and install
Min. working pressure: 1 bar
Depth: 710 mm
Connection: Inlet G1" female
Certifikácie: EN 15154-1/-2; ANSI Z358.1