Tlačiť
Safety cans
Dodávateľ: Köhler Technische Produkte
113-1241EA 116.24 EUR
113-1241 113-1242 113-1243
Safety cans
Nádoby bezpečnostní
Safety cans, made of high quality and chemically resistant stainless steel with PTFE gaskets. For safe and comfortable handling of small amounts of flammable liquids.
  • Easy handling, ergonomic design and high stability
  • Flame traps and explosion-proof
  • Grounding connection
  • Metallic flame barrier in filling/pouring opening
Certifikácie: ADR/RID/IMDG approval (UN number)