Tlačiť
Earmuffs, uvex x
Dodávateľ: Uvex
111-1573EA 50.68 EUR
111-1573
Earmuffs, uvex x
Chrániče mušlové Chrániče mušlové
The uvex x earmuffs provide protection against extreme high noise exposure levels. The small size and round design provides the security of perfect fitting, very high attenuation, low weight and comfortable pressure.
  • Round design combines light weight with a small capsule and high sound insulation
  • Foldable
  • Optimum protection in extremely noisy environments
SNR: 36 dB
Weight: 220 g
Certifikácie: EN 352-1