Tlačiť
Safety dispensing cans
Dodávateľ: asecos
MSGA14747EA 308.15 EUR
MSGA14747 MSGA14746 MSGA14745
Safety dispensing cans
Cans of high quality and chemically resistant stainless steel with PTFE gaskets. For safe and comfortable handling of small amounts of flammable liquids.
  • Easy handling, ergonomic design and high stability
  • Removable flame retarder made of perforated stainless steel
  • Fine dosing cap can be redacted through 360°
  • With ventilation an pressure relief valve