Tlačiť
Safety containers
Dodávateľ: asecos
MSGA2640EA 327.58 EUR
MSGA2640 MSGA11870
Safety containers
Nádoby bezpečnostní
Safety container ideal for corrosive liquids and flammables. Flame protection screen made of stainless steel. Grounding connection between container and liquid.
  • Sturdy construction
  • Operating elements and flame retarder made of stainless steel
  • Self-closing opening for pouring