Tlačiť
Jars, with screw cap
Dodávateľ: Saint-Gobain
216-3253EA 661.42 EUR
VERND1069029 VERND1069027 VERND1069025 VERND1069030
Jars, with screw cap
Straight sided jars, made of PFA, with screw caps.
  • Stackable
  • Marking area on the side of the jar for identification
  • Knurled tops for suregrip, easy opening and closing
These jars have very large openings and suitable for bulk solids, powders, pastes and digestion samples.