Tlačiť
ESD occupational shoes, lace-up, Uni6, 36762
Dodávateľ: Abeba
Uni6
ABEB36762/35EA 128.32 EUR
ABEB36762/35
ESD occupational shoes, lace-up, Uni6, 36762
Obuv
ESD shoes made from functional leather with honeycomb pattern and a TPU sole resistant to oils, fuels and animal fats.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • PU midsole
  • Slip resistant TPU sole
  • Water repellent and scratch proof
Colour: Blue
Certifikácie: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O1, FO, SRC