Chcete sa opäť prihlásiť?
Push-in caps for tubes
Dodávateľ: SEMADENI
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
Caps, Ø 3 mm 100 SEMA1435
Caps, Ø×H; 4×6 mm 100 SEMA1437
Caps, Ø×H; 5×6,5 mm 1.000 SEMA1439
Caps, Ø×H; 5,5×6,5 mm 100 SEMA1440 Blocked
Caps, Ø 6,5 mm 100 SEMA1442
Caps, Ø×H; 7×7,5 mm 100 SEMA1443
Caps, Ø×H; 8×7,5 mm 100 SEMA1445
Caps, Ø×H; 10×7,5 mm 100 SEMA1449 Blocked
Caps, Ø×H; 11×8 mm 100 215-0236 Blocked
Caps, Ø×H; 12,5×7,5 mm 100 SEMA7701
Caps, Ø×H; 13×8,5 mm 100 SEMA1455 Blocked
Caps, Ø×H; 13,5×8,5 mm 100 SEMA1456
Caps, Ø×H; 15×9 mm 100 SEMA1459 Blocked
Caps, Ø×H; 17×8,5 mm 100 SEMA1463 Blocked
Caps, Ø×H; 17,5×9 mm 100 SEMA1464 Blocked
Caps, Ø 19 mm 100 SEMA1467
Caps, Ø 22 mm 100 SEMA7702
Caps, Ø 29 mm 100 215-0216 Blocked
Caps, Ø×H; 25×10,5 mm 100 SEMA7706 Blocked
Caps, Ø×H; 26×13 mm 100 SEMA1481 Blocked
Caps, Ø×H; 29,5×12 mm 100 SEMA1488
Blocked Vaša organizácia zablokovala tento produkt. Prosím kontaktujte oddelenie nákupu pre viac informácii.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo