Chcete sa opäť prihlásiť?
Well plates, PP, WebSeal™, chromatography tested
Dodávateľ: Thermo Scientific
WebSeal
PP, non coated, non sterile, chromatography tested. Manufactured from ultra low bleed high purity basis resin and fully lot tested by GC/MS for organic extractables.
  • Microplates are chemically and thermally resistant and will tolerate temperatures from −80 °C to +121 °C
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • Ideal for pharmaceutical applications, sample collection and storage, combinatorial chemistry and HTS applications
  • U- and V- bottom well format for optimal sample recovery
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Tvar Výška Objem komôrky Odporúčaný pracovný objem Pk Číslo v katalógu VWR Množstvo
96-well PP MicroWell plate, round well V-shape well base, Ø 8 mm 14,4 mm 450 µl 10 - 400 µl 20 CRMA60180-P100
96-well PP MicroWell plate, round well V-shape well base, Ø 8 mm 14,4 mm 450 µl 10 - 400 µl 120 CRMA60180-P130
96-well PP MicroWell plate, round well U-shape well base, Ø 8 mm 14,5 mm 500 µl 20 - 450 µl 10 CRMA60180-P102
96-well PP MicroWell plate, round well U-shape well base, Ø 8 mm 14,5 mm 500 µl 20 - 450 µl 120 CRMA60180-P132
96-well PP MicroWell plate, round well U-shape well base, Ø 8 mm 31,6 mm 1300 µl 50 - 1000 µl 5 CRMA60180-P103
96-well PP MicroWell plate, round well U-shape well base, Ø 8 mm 31,6 mm 1300 µl 50 - 1000 µl 50 CRMA60180-P133
96-well PP MicroWell plate, round well U-shape well base, Ø 8 mm 44,0 mm 2000 µl 50 - 1900 µl 5 CRMA60180-P104
96-well PP MicroWell plate, round well U-shape well base, Ø 8 mm 44,0 mm 2000 µl 50 - 1900 µl 60 CRMA60180-P134
96-well PP plate, square well V-shape well base 44,0 mm 2000 µl 50 - 1900 µl 5 CRMA60180-P105
96-well PP plate, square well V-shape well base 44,0 mm 2000 µl 50 - 1900 µl 50 CRMA60180-P135
384-shallow well, standard height plate, square well U-shape well base 14,4 mm 58 µl 2 - 35 µl 100 CRMA60180-P136
384-well PP deep well plate, square well U-shape well base 22,0 mm 252 µl 5 - 240 µl 5 CRMA60180-P107
384-well PP deep well plate, square well U-shape well base 14,4 mm 120 µl 10 - 100 µl 20 CRMA60180-P108
384-well PP deep well plate, square well U-shape well base 14,4 mm 120 µl 10 - 100 µl 120 CRMA60180-P138
384-well PP deep well plate, square well V-shape well base 14,4 mm 145 µl 4 - 120 µl 10 CRMA60180-P109
384-well PP deep well plate, square well V-shape well base 14,4 mm 145 µl 4 - 120 µl 80 CRMA60180-P139
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
DOPLNKY
Rozšíriť všetko / Schovať všetko
Accessories
Opis Teplot. rozsah Pk Číslo v katalógu VWR Množstvo
Silicone sealing mat, solvent resistant, pre-slit, 96 round well, Ø 8 mm, clear - 10 CRMA60180-M100
EVA sealing mat, alcohol resistant, 96 round well, Ø 8 mm, clear - 5 CRMA60180-M101
Silicone/PTFE sealing mat, solvent resistant, 96 round well, dome base, Ø 8 mm, clear - 5 CRMA60180-M102
Silicone/PTFE sealing mat, solvent resistant, pre-slit, 96 round well, dome base, Ø 8 mm, clear - 5 CRMA60180-M103
Sealing mat applicator, handheld - 1 548-1895
Plate sealing tape, PE/silicone, clear −40...+90 °C 100 CRMA60180-M142
Plate sealing tape, PES/silicone, clear −70...+100 °C 100 CRMA60180-M143
Plate sealing tape, aluminium/silicone, silver –80...+120  °C 100 CRMA60180-M144
Plate sealing tape with synthetic rubber adhesive, polyolefin/rubber, clear - 100 CRMA60180-M145
Plate sealing tape with 3 layers, no adhesive at cavity area, PET/silicone/PET, clear - 100 CRMA60180-M146
Sealing tape applicator, handheld - 2 CRMA60180-M950
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo