Tlačiť
ESD safety ankle boots, lace-up, Anatom, 32168
Dodávateľ: Abeba
Anatom
ABEB32168/40EA 126.33 EUR
ABEB32168/40 ABEB32168/50 ABEB32168/42 ABEB32168/41 ABEB32168/37 ABEB32168/48 ABEB32168/47 ABEB32168/36 ABEB32168/39 ABEB32168/49 ABEB32168/38 ABEB32168/44 ABEB32168/43 ABEB32168/46 ABEB32168/45
ESD safety ankle boots, lace-up, Anatom, 32168
ESD ankle boots made from smooth leather, reflective material and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Sanitized® treatment reduces odour and bacterial growth
  • Protective steel toecap (200 J)
  • Scratch resistant coating in heel and toe area
Colour: Black
Certifikácie: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2, SRC.