Tlačiť
ESD occupational shoes, lace-up, Reflexor®, 36910
Dodávateľ: Abeba
Reflexor®
ABEB36910/36EA 141.78 EUR
ABEB36910/36 ABEB36910/39 ABEB36910/37 ABEB36910/38 ABEB36910/42 ABEB36910/40 ABEB36910/41
ESD occupational shoes, lace-up, Reflexor®, 36910
ESD clog made from smooth leather, respiration absorbing lining, and a fuel resistant PU sole.
  • Two replaceable Reflexor® integrated insoles (a massage integrated insole and a smooth comfort integrated insole)
  • Orthopaedic insoles possible
  • Extremely light, slip-resistant PU sole
Colour: Black
Certifikácie: EN 61340 (ESD); EN ISO 20347:2012; O1, SRC