Tlačiť
Occupational shoes, clogs, Arrow, 8600
Dodávateľ: Abeba
ABEB8600/40EA 82.3 EUR
ABEB8600/40 ABEB8600/39 ABEB8600/38 ABEB8600/46 ABEB8600/45 ABEB8600/37 ABEB8600/47 ABEB8600/36 ABEB8600/42 ABEB8600/41 ABEB8600/44 ABEB8600/43
Occupational shoes, clogs, Arrow, 8600
Anitstatic clogs made from smooth leather upper with leather heel patch and non foldable, adjustable heel strap. Ideal for use in kitchens.
  • Leather lining
  • Sanitized® treatment reduces odour and bacterial growth
  • Shock-absorbent PU midsole
  • Slip resistant PU sole
  • Adjustable instep area
Colour: White
Certifikácie: EN ISO 20347:2012; OB, A, E, FO, SRA