Tlačiť
Filter tips, sterile, ep Dualfilter T.I.P.S® SealMax
Dodávateľ: Eppendorf
epT.I.P.S® Dualfilter
613-2958 613-2959 613-2961
Filter tips, sterile, ep Dualfilter T.I.P.S® SealMax
Špičky pipetovací
These self-sealing tips with two-phase filter combine the same level of aerosol and biomolecule protection as ep Dualfilter T.I.P.S® with the reliable capability of sealing against liquid penetration upon contact.
  • Two-phase filter: Violet layer blocks liquids and binds aerosols; white layer repels drops and splashes and binds aerosols
  • Maximum protection against aerosols and biomolecules for pipette and sample
  • Made from pure, non recycled materials under cleanroom conditions without the use of biocides or plasticisers
  • PCR clean, sterile, pyrogen-free
The tips are ideal for methods where high sensitivity and reproducibility are essential. Perfect for applications such as PCR and microbiology where preserving sample integrity and preventing cross-contamination are critical.