Tlačiť
Cleanroom anti-fatigue mats, Ergomat® Nitril F
Dodávateľ: Ergomat
ERGONITRILF-BLACKEA 256.38 EUR
ERGONITRILF-BLACK
Cleanroom anti-fatigue mats, Ergomat® Nitril F
The Ergomat® Nitril F is made from nitrile rubber and PVC and is designed to increase comfort while reducing trip hazards, body pain, quality issues, and sick leave. It is ideal for areas where heat, oil and petrochemical resistance is crucial.
  • Upper side with cones
  • Fire retardant, withstands 600 °C (welding)
  • Can be autoclaved (610×915 mm only) and features great anti-bacterial cleaning properties
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin material guarantee durability and ergonomic benefits
  • Free from silicone and latex
Suitable for use in Class ISO 5 (100) cleanrooms.
Colour: Black
Certifikácie: Meets EN ISO/USDA/CFIA standards