Tlačiť
Microwave digestion vessels, Discover SP-D
Dodávateľ: CEM
CEMA927132 CEMA927130
Microwave digestion vessels, Discover SP-D
Systémy rozkladové Mineralizátory
Discover SP-D is a sequential, pressurised microwave digestion system that processes samples in 10 minutes or less, including cool down. The PowerMAX™ feature makes it possible for the Discover SP-D to introduce a maximum amount of energy to the sample while simultaneously cooling it.
  • Offers automated sample handling using Explorer automation deck and load methods in Synergy-D software
  • Available as single place system or with 48, 72 and 96 position decks with 10 or 35 ml vials
  • Processes around 6 samples per hour and up to 144 samples per day (up to 0,3 g samples)
  • ActiVent™ technology actively measures pressure and controls venting of the sample vial to prevent loss of elements, including volatiles such as mercury and arsenic
Focused™ microwave cavity, a self-tuning, single-mode design that produces a highly defined, extremely intense, microwave field that is very reproducible, a characteristic not normally associated with microwave cavities, as multi-mode cavities are widely known to have hot spots. This cavity design intensifies the coupling with acids and enables it to process samples much faster than multi-mode systems, without the need for turntables or diffusers.