Tlačiť
Glass tube/rod cutting knife, Kingcut
Dodávateľ: Jencons
JENC349-090
Glass tube/rod cutting knife, Kingcut
Nože a řezačky
Tungsten carbide blade, brass holder.
A double-edged reversible blade held firmly in a brass holder. If the edge is not in use it is protected within the handle. The Kingcut blade is made from tungesten carbide for use with borosilicate glass.