Tlačiť
Ion exchange column kit, MAb charge variant analysis
Dodávateľ: Thermo Scientific
555-7475EA 2331.05 EUR
HYPE076196
Ion exchange column kit, MAb charge variant analysis
MAb Charge Variant Analysis IEX Column Kit includes two ion exchange (IEX) specialty columns for MAb charge variants analysis. This kit is a convenient starter kit for researchers at the beginning of MAb analysis projects, and facilitates the screening of two columns for determination of the best column for their specific monoclonal antibody sample. The kit includes:
ProPac™ WCX-10 analytical column, 4×250 mm (HYPE054993), a weak cation exchange column, the industry standard for high-resolution, high-efficiency analysis of monoclonal antibodies and associated variants.
MAbPac™ SCX-10 analytical column, 4×250 mm (HYPE074625), a strong cation exchange column designed specifically for high-resolution, high-efficiency analysis of monoclonal antibodies and associated variants.