Tlačiť
No maintenance hydrogen generators
Dodávateľ: Perkin Elmer
PERCN9307073 PERCN9307072 PERCN9307071 PERCN9307074
No maintenance hydrogen generators
Generátory plynu Generátory vodíku
These hydrogen generators use membrane technology for electrolytic production of pure hydrogen gas, including exclusive no maintenance auto drying technology. They are ideal as a feed gas for all GC detectors. Hydrogen purity: >99,9999%.
  • No maintenance auto drying technology
  • Auto shut off for leak and water off procedure feature for safety
  • Audible alarms for variable operating conditions
  • 250 and 500 ml/min models are designed with cascading capability up to ten units
Delivery pressure: 10,6 bar