Tlačiť
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)
Dodávateľ: G Bioscienes
BC91EA 411.78 EUR
BC91
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)
Sulpho NHS acetate is a small, water-soluble compound used for blocking primary amines by acylation. It covalent bonds with amine groups to prevent specific interactions and conjugations. It is water-soluble and reacts at pH 7 to 9.
Vzorec: C₄H₄NNaO₆S
Mw: 259,17 g/mol
Číslo MDL: MFCD01861959
Číslo CAS: 152305-87-8
https://sk.vwr.com/stibo/web/std.lang.all/44/37/23914437.jpg