Tlačiť
Immobilised monomeric avidin for biotin purification
Dodávateľ: G-Biosciences
786-596EA 1237.89 EUR
GENO786-596
Immobilised monomeric avidin for biotin purification
Immobilised monomeric avidin resin is designed for the simple affinity chromatography purification of proteins, antibodies and other molecules with a biotin tag.
  • Minimal leaching
  • Non denaturing
  • Reusable
  • High specificity
Binding capacity: 1,2 mg biotinylated BSA/ml resin
Informace o dodávce: Available as a 50% resin slurry or as a complete kit containing a reusable monomeric avidin column and the respective buffers for successful purification of biotinylated molecules.