Tlačiť
Fluorescent nucleotides, CyDye™
Dodávateľ: Cytiva
PA55032EA 2944.15 EUR
PA55032 PA55031 PA53031
Fluorescent nucleotides, CyDye™
Činidla pro Microarray
CyDye®'s are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy® dye labelled nucleotides exhibit the following high sensitivity, low non specific binding, high photostability, high water solubility and pH insensitivity. Cy® Dye labelled nucleotides are available in a range of bright, intense colours with narrow emission bands, well suited for multiplexing within a single sample.
Applications: For labelling DNA and RNA probes, especially for multicolour analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.