Tlačiť
Earmuffs, Leightning® L0
Dodávateľ: Howard Leight by Honeywell
DALL1013460
Earmuffs, Leightning® L0
These ultra slim earmuffs with neckband with low attenuation provide reliable protection for marginal noise or intermittent exposure.
  • Neckband allow usage together with most welding helmets
  • Easy storage when not in use
  • Extremely lightweight design rests comfortably on the head
  • Air Flow Control™ Technology provides better, consistent attenuation, especially at low frequencies
  • Steel wire construction and robust design withstand demanding use, especially in tough environments
  • Snap-In ear cushions makes cushion replacement quick and easy
Earmuffs be used in most noise environments (between 100 dB to less than 105 dB(A)).
SNR: 22 dB
Weight: 132,5 g
Certifikácie: EN 352-1