Tlačiť
U-tube capillary viscometers, BS/U
Dodávateľ: Fungilab
FUNGCV006-105EA 134.88 EUR
FUNGCV006-105 FUNGCV006-106 FUNGCV006-107 FUNGCV006-108 FUNGCV006-101 FUNGCV006-102 FUNGCV006-103 FUNGCV006-104
U-tube capillary viscometers, BS/U
Viskozimetry Viskozimetry kapilární
Capillary viscometers with indelible timing marks and reverse flow for opaque liquids.
  • Constant value at 40 and 100 °C
  • Precision: ±0,3%
Liquid is drawn into the upper bulb by suction, then flowed down through the capillary into the lower bulb.
Certifikácie: ASTM D-445, D446, ISO 3104, ISO 3105 and UNE 400313
Balenie: Supplied with the calibration certificate issued in accordance with the conditions of accreditation granted by a National Metrological Register, the test uncertainty and its traceability to international standards UKAS.