Tlačiť
Heating mantles, WHLG2 series
Dodávateľ: Winkler
Pilz
WINKWHLG2E000100RC WINKWHLG2E002000RC WINKWHLG2E001000RC WINKWHLG2E000500RC WINKWHLG2E003000RC WINKWHLG2E000250RC WINKWHLG2E000050RC
Heating mantles, WHLG2 series
Ohřívače Hnízda topná
For round bottomed flasks. Mantles have crocheted outer jacket made from glass silk.
  • Maximum temperature 450 °C
  • Mantles for 50 and 100 ml flasks have one heating zone, larger units have two heating zones depending on model
  • Detachable plug-in connection (cable length 0,5 m)
  • RCD personal safety plug with 1 m cable