Tlačiť
Assay plates, Eppendorf® Assay/Reader Plates
Dodávateľ: Eppendorf
737-0227EA 975.55 EUR
737-0227 737-0249 737-0248 737-0239 737-0228 737-0171 737-0170 737-0172 737-0169
Assay plates, Eppendorf® Assay/Reader Plates
Mikrodestičky Mikrodestičky pro HTS
Eppendorf® Assay/Reader Plates are optimised for measuring absorbance assays in the visible and UV range, fluorescence and chemiluminescence assays. Solid plates are suitable for top-reading plate readers. Clear plates and black/clear plates can be used with top- and bottom-reading plate readers.
  • Clear VIS and UV-VIS plates for all absorbance assays in the visible and UV range
  • Black and white plates for fluorescence and luminescence detection
  • White plates optimised for highest sensitivity in the detection of luminescence signals by maximising reflection
  • PCR clean or sterile: Every lot is individually tested and certified for guaranteed purity and highest reliability
All Eppendorf microplates are manufactured in ISO certified clean room conditions. They are available in the purity levels PCR clean (free of human DNA, DNase, RNase and PCR inhibitors) or sterile. Each batch is tested and certified individually.