Tlačiť
Vortex mixer, peqTWIST
PEQL91-PTWIST
Vortex mixer, peqTWIST
Vortex mixer with powerful motor for mixing biological and chemical reagents.
  • Fits tubes 1,5 ml to 50 ml with standard head
  • Wide choice of adapters for a variety of tube sizes available separately