Tlačiť
Peqlab peqGOLD, Blotting Membranes, Nylon
Dodávateľ: Avantor
732-3198EA 535.83 EUR
PEQL39-2010
Peqlab peqGOLD, Blotting Membranes, Nylon
Nylon blotting membrane with high binding capacity for nucleic acids.
  • High material strength making it more suitable for repeated cycles of hybridising and 'stripping'
  • Stable against common solvents
  • Supplied on stable supporting layer
  • Pore diameter: 0,45 µm