Tlačiť
Orbital multi-tier shaker, Innova® 5000
Dodávateľ: New Brunswick
Innova®
NEWBM1196-0002EA 0 EUR
NEWBM1196-0002
Orbital multi-tier shaker, Innova® 5000
Třepačky Třepačky orbitální
Innova® 5000 Multi-tier open air shaker which handles hundreds of samples simultaneously even with all six fully loaded platforms and running at maximum speed. These six removable platforms accept flasks from 10 ml to 6 L.
  • Large agitation range provides versatility for culturing a wide variety of cell types
  • Counterbalanced orbit provides uniform agitation to all samples without being bolted to the floor
  • Digital LED display indicates speed, setpoint, running time and alarm conditions
  • Ergonomic design with cantilevered platforms provides unobstructed access to all flasks
This is designed for minimum stress and vibration of your samples. Remote monitoring and removable control module can be panel mounted or positioned remotely, fiber-optic link enables positioning controls up to 10 m. Ideal for use in an environmental room; cool-running brushless DC motor has no appreciable effect on chamber temperature.