Tlačiť
Rat plasma
Dodávateľ: Biowest
S2140-050EA 255.46 EUR
S2140-050 S2140-500
Rat plasma
Plazma
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.