Tlačiť
RNAPure™, peqGOLD
PEQL30-1010
RNAPure™, peqGOLD
Ready to use reagent for quick and simple purification of high quality total RNA. Contains phenol and guanidine isothiocyanate.
  • Suitable for small and large samples
  • Reliable and reproducible with high purity RNA