Tlačiť
Sample cutter (fork/knife), SP Bel-Art
Dodávateľ: Bel-Art
BELAF369660000EA 53.34 EUR
BELAF369660000
Sample cutter (fork/knife), SP Bel-Art
Nože a řezačky
PC, autoclavable. This molded one-piece tool consists of a rocker knife and a three tined fork on an easy to grip handle. It can be used for cutting and holding samples.
  • Ideal for use with foods and semi solid material
  • Dishwasher safe
  • Molded of rugged polycarbonate, autoclavable at 121 °C