Tlačiť
Desiccator, gas-ported, SP Bel-Art Techni-Dome®
Dodávateľ: Bel-Art Products, a part of SP
Techni-Dome®
BELAF420290001EA 1183.96 EUR
BELAF420290001
Desiccator, gas-ported, SP Bel-Art Techni-Dome®
Exsikátory Exsikátory plastové
Clear polycarbonate desiccator with large storage space, portable and easy to use. Two gas ports accept tubing (inner Ø 6,35 mm) and have easy turning PTFE stopcocks to facilitate the turbulence-free entrance of gases, minimising disturbance of the contents.
  • Interior volume of 65 L, holds 101 cans
  • Two removable PP shelves with Ø 365 and 225 mm
  • Side handles enable portability
  • Gas ports can be closed to allow use with standard desiccants
Designed to maintain a full vacuum (29" Hg, 737 mm) for twenty four hours at room temperature and stay gastight without a vacuum. For operation at temperatures down to 0 ºC, apply a light coating of vacuum grease to the gasket.