Tlačiť
pH liquid indicator
Dodávateľ: Industrial Physics
SIMESP3500EA 48.34 EUR
SIMESP3500
pH liquid indicator
This pH indicator spray can determine whether the surface of concrete is alkaline or acid and can be used in painting or repairing concrete.